Please login to your online account to display your discounted pricing

Promega™ Beetle Luciferin, Potassium Salts


Can be used as reporter genes for studying transcription regulation in transient assay systems and as markers for stably transformed eukaryotic cells

Manufacturer: promega™  E1605

 View more versions of this product

Catalog No. PRE1605

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 318.4
Quantity 1g
Format Anhydrous
Storage Requirements -70°C
Includes 1 x 1g Beetle Luciferin, Potassium salt