Please login to your online account to display your discounted pricing

Promega™ Beetle Luciferin, Potassium Salts DFS Item


Can be used as reporter genes for studying transcription regulation in transient assay systems and as markers for stably transformed eukaryotic cells

Manufacturer: promega™  E1603

 View more versions of this product

Catalog No. PR-E1603

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 250mg Beetle Luciferin, Potassium salt
Quantity 250mg
Storage Requirements -70°C
Format Anhydrous
Molecular Weight 318.4