Promega™ AttoPhos™ AP Fluorescent Substrate System DFS Item
A highly sensitive, flluorescent alkaline phosphatase substrate.
Manufacturer:  Promega™ S1001
 View more versions of this product
Catalog No. PR-S1001
Description
Description
  • Produces a low fluorescence signal until enzymatically acted upon, yielding detection of AP to 0.1 attomole
  • Provides low background
  • Offers linear kinetics over five orders of magnitude of AP concentration
  • Has an unusually large Stokes' shift (≈120nm), which leads to higher detection sensitivity
Specifications
Specifications
Crystalline Powder
AP Fluorescent
4°C
1 x 36mg AttoPhos substrate, 1 x 60mL AttoPhos buffer
1 x 36mg