Please login to your online account to display your discounted pricing

Promega™ Amino Acid Mixtures DFS Item


Compatible for use in the Flexi™ Lysate, TNT™ Lysate and standard Rabbit Reticulocyte Lysate Systems as well as in the Wheat Germ Extract and E. coli S30 Systems

Manufacturer: promega™  L9961

 View more versions of this product

Catalog No. PR-L9961

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -70°C
Quantity 175μL
Solution Type Amino Acid Mixtures
Format Amino Acid Mixture minus Methionine