Please login to your online account to display your discounted pricing

Promega™ Alkaline Phosphatase, Calf Intestinal DFS Item


Enzymes are available in convenient, cost-effective concentrations that are economical and easy to pipet

Manufacturer: promega™  M1821

 View more versions of this product

Catalog No. PR-M1821

  • / Each
 

Description & Specifications

Specifications

Concentration 1U/μL
Storage Buffer 10mM Tris HCl (pH 8), 1mM MgCl2, 0.1mM ZnCl2, 50mM KCl, 50% Glycerol
For Use With (Application) Prevent recircularization and religation of linearized cloning vector DNA by removing Phosphate groups from both 5'-termini, dephosphorylation of 5' Phosphorylated ends of DNA or RNA for subsequent labeling with (32P)ATP and T4 Polynucleotide Kinase
Color Colorless
Includes 10X Reaction Buffer: 0.5M Tris HCl (pH 9.3 at 25°C), 10mM MgCl2, 1mM ZnCl2, 10mM Spermidine
Quantity 1000U
Format Liquid
Source Calf intestinal mucosa
pH 8
Storage Requirements -20°C
Product Name Alkaline Phosphatase