missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ ABL1 Kinase Enzyme System DFS Item

Easily screen and profile ABL1 kinase inhibitors.

$602.41 - $602.41

Specifications

Format Fluid
Molecular Weight 135
Storage Requirements -70°C
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
PRV1901 View Documents Promega™
V1901
Cell differentiation, Cell division, Cell adhesion and stress response 10μg Each for $602.41
Description

Description

  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

Fluid
135
-70°C