Please login to your online account to display your discounted pricing

Alfa Aesar™ Prolactin-Releasing Peptide (12-31), Rat


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66882MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66882MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1mg
Format Solid
Sequence Thr-Pro-Asp-Ile-Asn-Pro-Ala-Trp-Tyr-Thr-Gly-Arg-Gly-Ile-Arg-Pro-Val-Gly-Arg-Phe-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Common Name PrRP-20, rat
Product Type Prolactin-Releasing Peptide (12-31), rat
Molecular Weight 2272.60
Purification Method HPLC
Cross Reactivity Rat