missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Prolactin Receptor Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Prolactin Receptor Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1338P1ABX

 View more versions of this product

Catalog No. MS1338P1ABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Prolactin Receptor Ab-1, Mouse Monoclonal Antibody.

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor) which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

Host Species: Mouse

Clone: B6.2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human prolactin receptor

Molecular Weight: ∽70kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Prolactin Receptor Ab-1
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG1
RUO
Primary
Human
Immunohistochemistry (Paraffin)
B6.2
Unlabeled
Concentrated
Semi-purified human prolactin receptor
100μL
Steroids Hormones and Growth Factors
Monoclonal