Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Prolactin Receptor Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Prolactin Receptor Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1338PABX

 View more versions of this product

Catalog No. MS1338PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Prolactin Receptor Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone B6.2
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Semi-purified human prolactin receptor
Isotype IgG1
Quantity 200μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Provide accurate, reproducible results with the Thermo Scientific™ Prolactin Receptor Ab-1, Mouse Monoclonal Antibody.

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor) which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

Host Species: Mouse

Clone: B6.2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human prolactin receptor

Molecular Weight: ∽70kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO