missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Prolactin Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Prolactin Ab-2, Mouse Monoclonal Antibody.

$297.27 - $1,472.78

Specifications

Antigen Prolactin Ab-2
Clone PRL02
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1362P View Documents Thermo Scientific™
MS1362P
Purified Ab with BSA and Azide 1mL Each for $1,250.69
 
MS1362P0 View Documents Thermo Scientific™
MS1362P0
Purified Ab with BSA and Azide 100μL Each for $297.27
 
MS1362P1 View Documents Thermo Scientific™
MS1362P1
Purified Ab with BSA and Azide 500μL Each for $819.82
 
MS1362P1ABX View Documents Thermo Scientific™
MS1362P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $1,022.53
 
MS1362PABX View Documents Thermo Scientific™
MS1362PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,472.78
 
MS1362R7 View Documents Thermo Scientific™
MS1362R7
Ready-to-use for IHC (UVLP) 7mL Each for $742.53
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Prolactin Ab-2, Mouse Monoclonal Antibody.

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. Prolactin producing cells make up approximately 20% of the pituitary.

Host Species: Mouse

Clone: PRL02

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human prolactin

Molecular Weight: 23.5kDa

Positive Control: Pituitary gland

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Prolactin Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG2a kappa
PRL02
Monoclonal
Murine
Steroids Hormones and Growth Factors
Human prolactin