Please login to your online account to display your discounted pricing
Science First™ Project Star Diffraction Grating
Manufacturer: science first™  PS08D
 View more versions of this product
Catalog No. S42544-4
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Overhead projector
Product Type Diffraction grating overhead
Includes Four 5cm square monochromatic filters (red, blue, green, yellow)