Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor, Rabbit Polyclonal Antibody.

$261.80 - $1,322.61

Specifications

Antigen Progesterone Receptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Format Quantity Price Quantity    

RB9017P

 
thermo scientific™ lab vision™
RB9017P
Concentrated 1mL Each for $1,322.61

RB9017P0

 
thermo scientific™ lab vision™
RB9017P0
Concentrated 100µL Each for $261.80

RB9017P1

 
thermo scientific™ lab vision™
RB9017P1
Concentrated 500µL Each for $734.24

RB9017R7

 
thermo scientific™ lab vision™
RB9017R7
Prediluted 7mL Each for $633.80
Description & Specifications

Specifications

Antigen Progesterone Receptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Recombinant protein encoding human progesterone receptor 412-526 aa
Isotype IgG

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor, Rabbit Polyclonal Antibody.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: 412-526aa

Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa

Molecular Weight: 81 and 116kDa

Positive Control: T47D cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not tested)

Note:

USA: RUO; Int'l: RUO