Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor, Rabbit Monoclonal Antibody

Progesterone Receptor, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

$500.59 - $2,586.35

Specifications

Antigen Progesterone Receptor (SP2)
Clone SP2
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RM9102R7

 
thermo scientific™ lab vision™
RM9102R7
Ready-to-use for IHC (UVLP) 7mL Each for $1,155.84

RM9102S

 
thermo scientific™ lab vision™
RM9102S
Culture Supernatant with Azide 1mL Each for $2,586.35

RM9102S0

 
thermo scientific™ lab vision™
RM9102S0
Culture Supernatant with Azide 100µL Each for $500.59

RM9102S1

 
thermo scientific™ lab vision™
RM9102S1
Culture Supernatant with Azide 500µL Each for $1,846.24
Description & Specifications

Specifications

Antigen Progesterone Receptor (SP2)
Clone SP2
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Recombinant protein encoding human progesterone receptor 412-526 aa

Employ part of a comprehensive antibody portfolio with the Thermo Scientific™ Progesterone Receptor, Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

The progesterone receptor (PgR) is an estrogen-regulated protein. It has been proposed that expression of PgR determination indicates a responsive estrogen receptor (ER) pathway, and therefore, may predict likely response to endocrine therapy in human breast cancer. A number of studies have shown that PgR determination provides supplementary information to ER, both in predicting response to endocrine therapy and estimating survival. PgR has proved superior to ER as a prognostic indicator in some studies.

Host Species: Rabbit

Clone: SP2

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: 412-526aa

Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa

Molecular Weight: 116 + 81kDa

Positive Control: Breast CA, T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting