Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-7, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-7, Mouse Monoclonal Antibody.

$303.06 - $1,441.16

Specifications

Antigen Progesterone Receptor Ab-7
Clone hPRa 7
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS197P

 
thermo scientific™ lab vision™
MS197P
200μg/mL 1mL Each for $1,401.33

MS197P0

 
thermo scientific™ lab vision™
MS197P0
200μg/mL 100µL Each for $303.06

MS197P1

 
thermo scientific™ lab vision™
MS197P1
200μg/mL 500µL Each for $955.63

MS197P1ABX

 
thermo scientific™ lab vision™
MS197P1ABX
1mg/mL 100µL Each for $1,017.57

MS197PABX

 
thermo scientific™ lab vision™
MS197PABX
1mg/mL 200µL Each for $1,441.16
Description & Specifications

Specifications

Antigen Progesterone Receptor Ab-7
Clone hPRa 7
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-7, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 7

Isotype: IgG1/κ

Species Reactivity: Human, Baboon, Monkey, Mouse, and Rat. Does not react with guinea pig. Others not known.

Epitope: N-terminal half of human PgR

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO