Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-7, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-7, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS197P1ABX

 View more versions of this product

Catalog No. MS197P1ABX

  • / Each
 

Description & Specifications

Specifications

Immunogen PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Format Concentrated
Quantity 100µL
Antigen Progesterone Receptor Ab-7
Isotype IgG2a kappa
Clone hPRa 7
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Host Species Mouse
Research Discipline Cancer and Tumor Biology
Classification Monoclonal
Conjugate Unlabeled
Type Primary
Concentration 1mg/mL

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-7, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 7

Isotype: IgG1/κ

Species Reactivity: Human, Baboon, Monkey, Mouse, and Rat. Does not react with guinea pig. Others not known.

Epitope: N-terminal half of human PgR

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO