missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody.

$374.10 - $1,803.04

Specifications

Antigen Progesterone Receptor Ab-6
Clone hPRa 6
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS196P View Documents Thermo Scientific™
MS196P
200μg/mL 1mL Each for $1,731.39
 
MS196P0 View Documents Thermo Scientific™
MS196P0
200μg/mL 100μL Each for $374.10
 
MS196P1 View Documents Thermo Scientific™
MS196P1
200μg/mL 500μL Each for $1,188.18
 
MS196P1ABX View Documents Thermo Scientific™
MS196P1ABX
1mg/mL 100μL Each for $1,290.00
 
MS196PABX View Documents Thermo Scientific™
MS196PABX
1mg/mL 200μL Each for $1,803.04
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 6

Isotype: IgG2b

Species Reactivity: Human, Cow, Rabbit, and Chicken. Does not react with monkey. Others not known.

Epitope: Not determined

Immunogen: Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form)

Positive Control: T47D cells

Cellular Localization: Nuclear Ab-6 preferentially reacts with B-form of progesterone receptor.

Recommended for:

  • Western Blotting (Not optimum)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Progesterone Receptor Ab-6
Immunofluorescence, Immunoprecipitation
Unconjugated
Murine
Cancer and Tumor Biology
Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
hPRa 6
Monoclonal
Concentrated
RUO
Primary
IgG2b