missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Progesterone Receptor Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-2, Mouse Monoclonal Antibody.

$1,322.31 - $1,322.31

Specifications

Antigen Progesterone Receptor Ab-2
Clone hPRa 2
Applications Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS192P Thermo Scientific
MS192P
Purified Ab with BSA and Azide 1mL Each for $1,322.31
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-2, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 2

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Horse, Rabbit, and Chicken. Does not react with monkey. Others not known.

Epitope: N-terminal half of human progesterone

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form)

Positive Control: T47D cells or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Progesterone Receptor Ab-2
Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
Unlabeled
Murine
Cancer and Tumor Biology
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
hPRa 2
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa