missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Progesterone Receptor Ab-2, Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-2, Mouse Monoclonal Antibody.

Supplier:  Thermo Fisher Scientific MS192P

 View more versions of this product

Catalog No. MS192P



Description

Description

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 2

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Horse, Rabbit, and Chicken. Does not react with monkey. Others not known.

Epitope: N-terminal half of human progesterone

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form)

Positive Control: T47D cells or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Progesterone Receptor Ab-2
Monoclonal
200μg/mL
Mouse
1 mL
Cancer and Tumor Biology
Bovine, Chicken, Equine, Human, Rabbit
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
hPRa 2
Unconjugated
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
RUO
Primary
Concentrated
SDS
Documents

Documents

Promotions

Promotions