Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody. 

$680.29 - $3,182.27

Specifications

Antigen Progesterone Receptor Ab-14
Clone PgR636
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1882R7

 
thermo scientific™ lab vision™
MS1882R7
7mL Each for $1,560.55

MS1882S

 
thermo scientific™ lab vision™
MS1882S
1mL Each for $3,182.27

MS1882S0

 
thermo scientific™ lab vision™
MS1882S0
100μL Each for $680.29

MS1882S1

 
thermo scientific
MS1882S1
500μL Each for $2,328.92
Description & Specifications

Specifications

Antigen Progesterone Receptor Ab-14
Clone PgR636
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Formalin fixed recombinant full length A-form of human progesterone receptor
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: PgR636

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal (shared by A- and B-form)

Immunogen: Formalin fixed recombinant full-length A-form of human progesterone receptor

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO