Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody.

$280.10 - $1,398.67

Specifications

Antigen Progesterone Receptor Ab-13
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1492P

 
thermo scientific™ lab vision™
RB1492P
Purified Ab with BSA and Azide 1mL Each for $1,297.47

RB1492P0

 
thermo scientific™ lab vision™
RB1492P0
Purified Ab with BSA and Azide 100μL Each for $280.10

RB1492P1

 
thermo scientific™ lab vision™
RB1492P1
Purified Ab with BSA and Azide 500μL Each for $776.86

RB1492P1ABX

 
thermo scientific™ lab vision™
RB1492P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $857.47

RB1492PABX

 
thermo scientific™ lab vision™
RB1492PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,398.67
Description & Specifications

Specifications

Antigen Progesterone Receptor Ab-13
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen A synthetic peptide derived from C-terminus of human progesterone receptor protein
Isotype IgG

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-terminus of human progesterone receptor protein

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO