Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1492PABX

 View more versions of this product

Catalog No. RB1492PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Progesterone Receptor Ab-13
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic peptide derived from C-terminus of human progesterone receptor protein
Isotype IgG
Quantity 1mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human
Target Species Murine

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-terminus of human progesterone receptor protein

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO