Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1492P1ABX

 View more versions of this product

Catalog No. RB1492P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Host Species Rabbit
Isotype IgG
Quantity 500µL
Antigen Progesterone Receptor Ab-13
Cross Reactivity Human
Cross Reactivity Mouse
Applications Western Blotting
Immunogen A synthetic peptide derived from C-terminus of human progesterone receptor protein
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Conjugate Unlabeled
Concentration 1mg/mL
Research Discipline Cancer and Tumor Biology
Classification Polyclonal

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-13, Rabbit Polyclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-terminus of human progesterone receptor protein

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO