Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Ab-1, Mouse Monoclonal Antibody.

$242.45 - $693.88

Specifications

Antigen Progesterone Ab-1
Clone 4P01; same as 1E11
Applications Enzyme Linked Immunosorbent Assay
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1040P

 
thermo scientific™ lab vision™
MS1040P
200μg/mL 1mL Each for $607.73

MS1040P0

 
thermo scientific™ lab vision™
MS1040P0
200μg/mL 100µL Each for $242.45

MS1040P1

 
thermo scientific™ lab vision™
MS1040P1
200μg/mL 500µL Each for $417.59

MS1040P1ABX

 
thermo scientific™ lab vision™
MS1040P1ABX
1mg/mL 100µL Each for $543.55

MS1040PABX

 
thermo scientific™ lab vision™
MS1040PABX
1mg/mL 200µL Each for $693.88
Description & Specifications

Specifications

Antigen Progesterone Ab-1
Clone 4P01; same as 1E11
Applications Enzyme Linked Immunosorbent Assay
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity All
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Progesterone-11alpha-hemisuccinate conjugated to BSA
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Ab-1, Mouse Monoclonal Antibody.

Progesterone is a steroid hormone secreted by luteinized theca cells of normal ovary. The determination of progesterone concentrations in the body fluids is of great value for endocrinological investigations in women.

Host Species: Mouse

Clone: 4P01; same as 1E11

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: Progesterone-11α-hemisuccinate conjugated to BSA

Positive Control: T47D, Luteinized theca cells of normal ovary

Cellular Localization: Cytoplasmic and secreted In competitive binding immunoassay, Ab-1 shows minimal cross-reaction with 17alpha-hydroxyprogesterone (0.8%); 3Beta-hydroxypregn-5-en-20-one (<0.1%); 20alpha-dihydroprogesterone (0.1%); testosterone (<0.1%); 17Beta-estradiol (<0.1%); and cortisol (<0.1%). Ab-1 may also prove useful in identification of ovarian tumors.

Recommended for:

  • ELISA
  • Radioimmunoassay (RIA)

Note:

USA: RUO; Int'l: RUO