Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Progesterone Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1040PABX

 View more versions of this product

Catalog No. MS1040PABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Immunogen Progesterone-11alpha-hemisuccinate conjugated to BSA
Cross Reactivity All
Antigen Progesterone Ab-1
Host Species Mouse
Applications Enzyme Linked Immunosorbent Assay
Isotype IgG2a kappa
Regulatory Status RUO
Quantity 200µL
Clone 4P01; same as 1E11
Conjugate Unlabeled
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Ab-1, Mouse Monoclonal Antibody.

Progesterone is a steroid hormone secreted by luteinized theca cells of normal ovary. The determination of progesterone concentrations in the body fluids is of great value for endocrinological investigations in women.

Host Species: Mouse

Clone: 4P01; same as 1E11

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: Progesterone-11α-hemisuccinate conjugated to BSA

Positive Control: T47D, Luteinized theca cells of normal ovary

Cellular Localization: Cytoplasmic and secreted In competitive binding immunoassay, Ab-1 shows minimal cross-reaction with 17alpha-hydroxyprogesterone (0.8%); 3Beta-hydroxypregn-5-en-20-one (<0.1%); 20alpha-dihydroprogesterone (0.1%); testosterone (<0.1%); 17Beta-estradiol (<0.1%); and cortisol (<0.1%). Ab-1 may also prove useful in identification of ovarian tumors.

Recommended for:

  • ELISA
  • Radioimmunoassay (RIA)

Note:

USA: RUO; Int'l: RUO