missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Productainer™ BPC Bag with 4 Top Ports

Using these BPC bags with 4 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

$328.11 - $384.98

Specifications

Description Three-dimensional disposable cell culture/bioprocess bag
Film Description CX3-9 Film
Port Detail 3/8in. Quick Disconnect Insert (MPC Series); 1/4in. Male Luer Lock; 1/4in. Female Luer Lock; 3/8in. Quick Disconnect Body (MPC Series)
Sterilization Method Gamma Irradiated
Storage Requirements Ambient
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
SH3065101 Thermo Scientific™
SH3065101
50L Each for $328.11
SH3065103 Thermo Scientific™
SH3065103
200L Each for $384.98
SH3065102 Thermo Scientific™
SH3065102
100L Each for $332.60
Specifications

Description

Using Thermo Scientific™ Productainer™ BPC bag with 4 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

  • Square tube design, 3-dimensional

  • Constructed with Thermo Scientific CX5-14 film with a contact surface composed of ultra-low density polyethylene
  • Four top ports (2 to 25.4mm (1in.); 2 to 12.7mm (1/2in.))
  • Line 1:

  • 9.5mm (3/8in.) MPC insert constructed of polycarbonate with 121.9cm (48in.) of C-Flex™ SEBS tubing (9.5mm (3/8in.) ID by 15.9mm
  • (5/8in.) OD); Variable length diptube: 50L - 38cm (15in.); 100L - 71cm (28in.); 200L - 81cm (32in.)
  • Line 2:

  • 6.4mm (1/4in.) Male Luer Lock constructed of polypropylene with 121.9cm (48in.) of C-Flex SEBS tubing (6.4mm (1/4in.) ID by 9.5mm (3/8in.) OD)
  • Line 3:

  • 6.4mm (1/4in.) Female Luer Lock constructed of polypropylene with 121.9cm (48in.) of C-Flex SEBS tubing (6.4mm (1/4in.) ID by 9.5mm (3/8in.) OD)
  • Line 4:

  • 9.5mm (3/8in.) MPC body constructed of polycarbonate with 121.9cm (48in.) of C-Flex SEBS tubing (9.5mm (3/8in.) ID by 15.9mm (5/8in.) OD);
  • Diptube length:
  • 30.5cm (12in.)
  • Designed to be used with flat bottom linear low density polyethylene drums with clamps and matching lids
  • Plastic drum dolly for ergonomic convenience
Description & Specifications

Specifications

Three-dimensional disposable cell culture/bioprocess bag
3/8in. Quick Disconnect Insert (MPC Series); 1/4in. Male Luer Lock; 1/4in. Female Luer Lock; 3/8in. Quick Disconnect Body (MPC Series)
Ambient
CX3-9 Film
Gamma Irradiated
No