missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Productainer™ BPC Bag with 2 Top Ports

Using these BPC bags with 2 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

$249.92 - $308.63

Specifications

Description Three-dimensional disposable cell culture/bioprocess bag; 100L; Two Top Ports: MPC Insert, MPC Body with 48 in. Dip Tube and 12 in. Dip Tube
Film Description CX3-9 Film
Port Detail 3/8in. Quick Disconnect Insert (MPC Series); 3/8in. Quick Disconnect Body (MPC Series)
Sterilization Method Gamma Irradiated
Storage Requirements Ambient
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
SH3064902 Thermo Scientific™
SH3064902
100L Each for $308.63
SH3064903 Thermo Scientific™
SH3064903
200L Each for $304.29
SH3064901 Thermo Scientific™
SH3064901
50L Each for $249.92
Specifications

Description

Using Thermo Scientific™ Productainer™ BPC bag with 2 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainer BPCs can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

Description & Specifications

Specifications

Three-dimensional disposable cell culture/bioprocess bag; 100L; Two Top Ports: MPC Insert, MPC Body with 48 in. Dip Tube and 12 in. Dip Tube
3/8in. Quick Disconnect Insert (MPC Series); 3/8in. Quick Disconnect Body (MPC Series)
Ambient
CX3-9 Film
Gamma Irradiated
No