missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Procion Blue H-5R

$62.80 - $209.34

Chemical Identifiers

CAS 12236-84-9
MDL Number MFCD00070851
Synonym C.I. 181575; Reactive Blue 13
Chemical Name or Material Procion Blue H-5R
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1271914 SDS Alfa Aesar™
A1271914
25g Each for $62.80
AAA1271922 SDS Alfa Aesar™
A1271922
100g Each for $209.34
Specifications Description & Specifications

Chemical Identifiers

12236-84-9
MFCD00070851
C.I. 181575; Reactive Blue 13
Procion Blue H-5R

Specifications

12236-84-9
C.I. 181575; Reactive Blue 13
Procion Blue H-5R
MFCD00070851
25g