missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Prochlorperazine Dimaleate, TCI America™

Manufacturer:  TCI America P236825G

 View more versions of this product

Catalog No. P236825G



Specifications

Specifications

Prochlorperazine Dimaleate
≥98.0% (HPLC,T)
MFCD00069330
DSKIOWHQLUWFLG-SPIKMXEPSA-N
(Z)-but-2-enedioic acid;2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
5281032
606.09
White-Yellow
25g
84-02-6
C28H32ClN3O8S
2-Chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine Dimaleate
CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)Cl.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O
606.087
CHEBI:8436
Crystal-Powder at 20°C
194°C

Chemical Identifiers

84-02-6
606.087
DSKIOWHQLUWFLG-SPIKMXEPSA-N
5281032
(Z)-but-2-enedioic acid;2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
C28H32ClN3O8S
MFCD00069330
2-Chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine Dimaleate
CHEBI:8436
CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)Cl.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : SO3150000

TSCA : No

Documents