missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Prochloraz 98.0+%, TCI America™
$746.56 - $746.56
Chemical Identifiers
CAS | 67747-09-5 |
---|---|
Molecular Formula | C15H16Cl3N3O2 |
Molecular Weight (g/mol) | 376.662 |
MDL Number | MFCD00078735 |
InChI Key | TVLSRXXIMLFWEO-UHFFFAOYSA-N |
Synonym | prochloraz, mirage, octave, prelude, ascurit, sporgon, sportak, mirage pesticide, dibavit, sprint |
PubChem CID | 73665 |
ChEBI | CHEBI:8434 |
IUPAC Name | N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]imidazole-1-carboxamide |
SMILES | CCCN(CCOC1=C(C=C(C=C1Cl)Cl)Cl)C(=O)N2C=CN=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
P21375G
|
TCI America
P21375G |
5 g |
N/A
|
|
|||||
Chemical Identifiers
67747-09-5 | |
376.662 | |
TVLSRXXIMLFWEO-UHFFFAOYSA-N | |
73665 | |
N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]imidazole-1-carboxamide |
C15H16Cl3N3O2 | |
MFCD00078735 | |
prochloraz, mirage, octave, prelude, ascurit, sporgon, sportak, mirage pesticide, dibavit, sprint | |
CHEBI:8434 | |
CCCN(CCOC1=C(C=C(C=C1Cl)Cl)Cl)C(=O)N2C=CN=C2 |
Specifications
67747-09-5 | |
C15H16Cl3N3O2 | |
5 g | |
prochloraz, mirage, octave, prelude, ascurit, sporgon, sportak, mirage pesticide, dibavit, sprint | |
CCCN(CCOC1=C(C=C(C=C1Cl)Cl)Cl)C(=O)N2C=CN=C2 | |
376.662 | |
CHEBI:8434 | |
≥98.0% (HPLC,N) | |
Prochloraz |
50°C | |
MFCD00078735 | |
3077 | |
TVLSRXXIMLFWEO-UHFFFAOYSA-N | |
N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]imidazole-1-carboxamide | |
73665 | |
376.66 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : NI4000400
TSCA : No
Recommended Storage : Refrigerator