Please login to your online account to display your discounted pricing

Procaterol hydrochloride, Tocris

Potent β2 agonist

$166.60 - $693.49

Chemical Identifiers

Chemical Name or Material Procaterol hydrochloride
Assay Percent Range >99%
Molecular Formula C16H22N2O3·HCl
Formula Weight 326.82
Synonym (±)-erythro-8-Hydroxy-5-[1-hydroxy-2-(isopropylamino)butyl]carbostyril hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

110250

 
r & d systems
RDS110250
50mg Each for $693.49

110210

 
r & d systems
RDS110210
10mg Each for $166.60
Description & Specifications

Specifications

Formula Weight 326.82
Quantity 50mg
Chemical Name or Material Procaterol hydrochloride
Assay Percent Range >99%
Molecular Formula C16H22N2O3·HCl
CAS 62929-91-3
Synonym (±)-erythro-8-Hydroxy-5-[1-hydroxy-2-(isopropylamino)butyl]carbostyril hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature