missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Procaine | |
62°C | |
C13H20N2O2 | |
5 g | |
MFDFERRIHVXMIY-UHFFFAOYSA-N | |
2-(diethylamino)ethyl 4-aminobenzoate | |
4914 | |
236.32 | |
Crystalline Powder |
59-46-1 | |
White | |
MFCD00007893 | |
procaine, vitamin h3, novocaine, spinocaine, duracaine, procain, nissocaine, novocain, 2-diethylamino ethyl 4-aminobenzoate, procaina | |
CCN(CC)CCOC(=O)C1=CC=C(C=C1)N | |
236.315 | |
CHEBI:8430 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
59-46-1 | |
236.315 | |
MFDFERRIHVXMIY-UHFFFAOYSA-N | |
4914 | |
2-(diethylamino)ethyl 4-aminobenzoate |
C13H20N2O2 | |
MFCD00007893 | |
procaine, vitamin h3, novocaine, spinocaine, duracaine, procain, nissocaine, novocain, 2-diethylamino ethyl 4-aminobenzoate, procaina | |
CHEBI:8430 | |
CCN(CC)CCOC(=O)C1=CC=C(C=C1)N |
Safety and Handling
EINECSNumber : (9)-0162
RTECSNumber : DG2100000
TSCA : No