Please login to your online account to display your discounted pricing

Procainamide hydrochloride, 99%, ACROS Organics™

$41.75 - $41.75

Chemical Identifiers

Chemical Name or Material Procainamide hydrochloride
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H21N3O·HCl
Linear Formula H2NC6H4CONHCH2CH2N(C2H5)2·HCl
Formula Weight 271.78
MDL Number MFCD00012998
Synonym 4-Amino-N-(2-diethylaminoethyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC207300250

  SDS
fisher scientific acros
207300250
Glass bottle 25g Each for $41.75
Description & Specifications

Specifications

Physical Form Powder
Assay ≥98.5%
Formula Weight 271.78
Quantity 25g
Color White to slightly yellow
Beilstein 14, III, 1077
Merck Index 15, 7874
Melting Point 166° to 170°C
Linear Formula H2NC6H4CONHCH2CH2N(C2H5)2·HCl
Packaging Glass bottle
Synonym 4-Amino-N-(2-diethylaminoethyl)benzamide
Chemical Name or Material Procainamide hydrochloride
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C13H21N3O·HCl
MDL Number MFCD00012998
CAS 614-39-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.