missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Probenecid, TCI America™

Manufacturer:  TCI America P197525G

 View more versions of this product

Catalog No. P197525G



Specifications

Specifications

Probenecid
200°C
57-66-9
MFCD00038402
DBABZHXKTCFAPX-UHFFFAOYSA-N
4-(dipropylsulfamoyl)benzoic acid
4911
285.36
≥98.0% (HPLC,T)
White
25g
C13H19NO4S
probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(=O)O
285.358
CHEBI:8426
Crystal-Powder at 20°C

Chemical Identifiers

57-66-9
285.358
DBABZHXKTCFAPX-UHFFFAOYSA-N
4911
4-(dipropylsulfamoyl)benzoic acid
C13H19NO4S
MFCD00038402
probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
CHEBI:8426
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : DG9400000

TSCA : Yes

Documents