Please login to your online account to display your discounted pricing

Alfa Aesar™ Probenecid, 98%

$74.28 - $223.00

Chemical Identifiers

Chemical Name or Material Probenecid
CAS 57-66-9
Molecular Formula C13H19NO4S
Formula Weight 285.36
MDL Number MFCD00038402
Merck Index 14,7754
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2001088

  SDS
alfa aesar
B2001088
2.5g Each for $74.28

AAB2001009

  SDS
alfa aesar™
B2001009
10g Each for $223.00
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P304+P340-P362-P312-P405-P403+P233-P501a
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Formula Weight 285.36
Quantity 2.5g
TSCA TSCA
Assay 98%
Health Hazard 1 Danger
Merck Index 14,7754
RTECS Number DG9400000
MDL Number MFCD00038402
Assay Percent Range 0.98
Chemical Name or Material Probenecid
CAS 57-66-9
Molecular Formula C13H19NO4S

Solubility
Soluble in water (partly miscible), solutions of alkali hydroxides,, ethanol (100 mM), acetone, chloroform, sodium hydroxide (1 M), DMSO (100 mM), methanol, dilute acids (practically insoluble), dilute alkalies, and ether (slightly soluble).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P304+P340-P362-P312-P405-P403+P233-P501a

GHS H Statement
H301-H315-H319-H335