missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Proadifen hydrochloride

Manufacturer:  Alfa Aesar™ J6383304

 View more versions of this product

Catalog No. AAJ6383304



Specifications

Specifications

Proadifen hydrochloride
122°C to 123°C
62-68-0
MFCD00055151
Soluble in methanol or water
CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl
389.964
389.97
White
2g
C23H32ClNO2
proadifen hcl
FHIKZROVIDCMJA-UHFFFAOYSA-N
2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
65341
Powder

Chemical Identifiers

62-68-0
389.964
FHIKZROVIDCMJA-UHFFFAOYSA-N
65341
CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl
C23H32ClNO2
MFCD00055151
proadifen hcl
2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YV7175000

Recommended Storage : 2° to 8°C

Documents