Please login to your online account to display your discounted pricing

Alfa Aesar™ [Pro&superscript_9;] Substance P

For cellular and molecular biology applications

Specifications

Product Type [Pro9] Substance P
Molecular Weight 1387.71
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Pro-Leu-Met-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66198EXD

 
alfa aesar™
J66198EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66198MCR

 
alfa aesar™
J66198MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Pro9] Substance P
Molecular Weight 1387.71
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Pro-Leu-Met-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC