PRKX Recombinant Human Protein DFS Item
PRKX Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3813
Catalog No. PV3813
Description
Description
PRKX is a serine⁄threonine kinase related to the catalytic subunit of the cAMP-dependent protein kinases.
Specifications
Specifications
NP_005035
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
68.4kDa
90% by SDS-PAGE
–80°C
1 Each
PRKX, PKX1
PRKX was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 1μM cAMP, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL kemptide peptide substrate (LRRASLG) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
PKA
440nmoles of phosphate transferred to Kemptide peptide substrate (LRRASLG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL
GST Tag
0.38mg/mL
3960, 5613
PRKX Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
PRKX Recombinant Human Protein
Kinase assay
Baculovirus
PRKX Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.