missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PRKG2 (PKG2) Recombinant Human Protein DFS Item

PRKG2 (PKG2) exhibits many cellular functions.

$2645.00 - $11260.00

Specifications

Accession Number NP_006250
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Formulation 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Molecular Weight (g/mol) 116.6kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR7572A DFS Item
View Documents Promotion Details
Invitrogen™
PR7572A
100μg
Each for $2,645.00
 
PV4277 DFS Item
View Documents Promotion Details
Invitrogen™
PV4277
1mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

PRKG2 (PKG2) has cAMP- and cGMP-dependent protein kinase activity.

Specifications

Specifications

NP_006250
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
PKG Kinase Family
Recombinant
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
116.6kDa
RUO - research use only
PRKG2 (PKG2)
Human
GST Tag
Frozen
SDS
missing translation for 'documents'

missing translation for 'documents'