Invitrogen™ PRKG2 (PKG2) Recombinant Human Protein DFS Item
PRKG2 (PKG2) exhibits many cellular functions.
Manufacturer:  Invitrogen™ LSPV4277
 View more versions of this product
Catalog No. PV4277
Description
Description
PRKG2 (PKG2) has cAMP- and cGMP-dependent protein kinase activity.
Specifications
Specifications
NP_006250
Frozen
116.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PKG Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PRKG2 (PKG2)
Baculovirus
Human
GST Tag
Documents