Invitrogen™ PRKD2 (PKD2) Recombinant Human Protein DFS Item
PRKD2 (PKD2) is a member of the protein kinase D serine/threonine protein kinase family.
Manufacturer:  Invitrogen™ LSPV4042
 View more versions of this product
Catalog No. PV4042
Description
Description
PRKD2 is a component in the signal transduction pathway induced by phorbol esters and the CCK b gastrin receptor.
Specifications
Specifications
NP_057541
Frozen
124.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PKD Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PRKD2 (PKD2)
Baculovirus
Human
GST Tag
Documents