Invitrogen™ PRKD1 (PKC mu) Recombinant Human Protein DFS Item
PRKC1 (PKC mu) is a serine/threonine protein kinase that is phospholipid-dependant and activated by diacylglycerol and phorbol ester TPA.
Manufacturer:  Invitrogen™ LSPV4041
 View more versions of this product
Catalog No. PV4041
Description
Description
PRKC1 behaves similarly to most PKC isoenzymes of the PKC family, however, PRKC1 differs in some structural and enzymatic features from the other PKC isotypes, and has been shown to be involved in the signal transduction of a wide range of biological responses.
Specifications
Specifications
NP_002733
Kinase Assay
Frozen
129.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
PKD Kinase Family
Yes
Baculovirus
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PRKD1 (PKC mu), PKD
Human
GST Tag
Documents