PRKCQ (PKC theta) Recombinant Human Protein DFS Item
PRKCQ (PKC theta)
Manufacturer:  Life Technologies LSPV3605
 View more versions of this product
Catalog No. PV3605
Description
Description
The PKC family of serine⁄threonine kinases, including PRKCQ (PKC theta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications
Specifications
NP_006248
Human
0.34mg/mL
Frozen
84.4kDa
90% by SDS-PAGE
–80°C
1 Each
PRKCQ (PKC theta) Recombinant Human Protein
PKC
3505nmoles of phosphate transferred to PKC epsilon peptide substrate (ERMRPRKRQGSVRRRV) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.5 to 2μg/mL
His Tag
Baculovirus
Kinase assay
Protein assays and analysis
Frozen
PRKCQ (PKC theta) Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
PRKCQ (PKC theta)
PRKCQ (PKC theta) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 0.1mM EGTA, 1X lipid mix (200μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 100μg/mL PKC epsilon peptide substrate (ERMRPRKRQGSVRRRV) and trace [32P]-γ-ATP for 10 minutes at 30°C
PRKCQ (PKC theta) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.