missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PRKCQ (PKC theta) Recombinant Human Protein DFS Item

PRKCQ (PKC theta)

Manufacturer:  Life Technologies LSP2996

 View more versions of this product

Catalog No. P2996



Description

Description

The PKC family of serine⁄threonine kinases, including PRKCQ (PKC theta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications

Specifications

NP_006248
Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 20% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
PRKCQ
Baculovirus
Full Length, Recombinant
Signal Transduction
Kinase Assay
84.4 kDa
PRKCQ (PKC theta)
His Tag
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Ser⁄Thr Kinases: AGC Group
LanthaScreen® Antibody, LanthaScreen® Substrate, Z'-LYTE™, Omnia® Bulk Substrate
protein kinase C, theta
Kinases & Inhibitors
Serine⁄Threonine Kinases
Human
5584, 5588
5μg
PKC Kinase Family
Documents

For Research Use Only. Not for use in diagnostic procedures.