missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PRKCB1 (PKC beta I) Recombinant Human Protein DFS Item

PRKCB1 (PKC beta I)

$449.00 - $8,010.00

Specifications

Accession Number NP_991100.1
Assays PRKCB1 (PKC beta I) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 100μM CaCl2, 1X lipid mix (100μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 200μg/mL histone H1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name PRKCB1 (PKC beta I) Recombinant Human Protein
Expression System Baculovirus
KinaseFamily PKC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P2291
View Documents
Life Technologies
LSP2291
5μg Each for $449.00
 
P2281
View Documents
Life Technologies
LSP2281
20μg Each for $980.00
 
PV3601
View Documents
Life Technologies
LSPV3601
1mg Each for $8,010.00
 
Description

Description

The PKC family of serine⁄threonine kinases, including PRKCB1 (PKC beta I), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications

Specifications

NP_991100.1
PRKCB1 (PKC beta I) Recombinant Human Protein
PKC
PRKCB1 (PKC beta I) Recombinant Human Protein
PRKCB1 (PKC beta I) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
None
PRKCB1 (PKC beta I) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 100μM CaCl2, 1X lipid mix (100μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 200μg/mL histone H1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
76.9kDa
1 Each
95% by SDS-PAGE
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.