Invitrogen™ PRKACB (PRKAC beta) Recombinant Human Protein DFS Item
The catalytic subunit C-beta of PKA (PRKACB) is a member of the Ser/Thr protein kinase family. Recombinant full-length human PRKACB was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Manufacturer:  Invitrogen™ LSPV6390
 View more versions of this product
Catalog No. PV6390
Description
Description
The PKA catalytic subunit consists of three gene products: C-alpha, C-beta, and C-gamma. The catalytic subunit C-beta has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
66.9kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Yes
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol
1mg
PKAb, cAPKb, MGC9320, MGC41879, DKFZp781I2452
Human
GST Tag
Documents