missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ PRKACB (PRKAC beta) Recombinant Human Protein
The catalytic subunit C-beta of PKA (PRKACB) is a member of the Ser/Thr protein kinase family. Recombinant full-length human PRKACB was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Manufacturer: Invitrogen™ LSPV6389
Description
The PKA catalytic subunit consists of three gene products: C-alpha, C-beta, and C-gamma. The catalytic subunit C-beta has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma | |
66.9kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Kinase Assay | |
Yes |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol | |
100μg | |
PKAb, cAPKb, MGC9320, MGC41879, DKFZp781I2452 | |
Human | |
GST Tag |