PRKACA (PKA) Recombinant Human Protein DFS Item
PRKACA (PKA) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4040
 View more versions of this product
Catalog No. PV4040
Description
Description
PRKACA (PKA) is a serine⁄threonine protein kinase that phosphorylates a broad spectrum of substrates. PRKACA regulates many cellular processes including cell growth, cell differentiation, ion-channel conductivity, gene transcription, and synaptic release of neurotransmitters.
Specifications
Specifications
NP_002721.1
Protein assays and analysis
Frozen
PRKACA (PKA) Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
PRKACA (PKA)
PRKACA (PKA) was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.4), 24mM MgCl2, 240μM ATP, 180μg/mL kemptide peptide substrate (LRRASLG) and trace [32P]-γ-ATP for 10 minutes at 30°C
E. coli
PRKACA (PKA) Recombinant Human Protein
Human
0.33mg/mL
Frozen
43.5kDa
90% by SDS-PAGE
–80°C
1 Each
PRKACA (PKA) Recombinant Human Protein
Kinase assay
PKA
4,800nmoles of phosphate transferred to Kemptide peptide substrate (LRRASLG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.417μg/mL
His Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.