Please login to your online account to display your discounted pricing

Tocris™ Pristimerin

Potent, reversible MAGL inhibitor

$257.85 - $1,073.10

Specifications

Product Type Pristimerin
Inhibitors Pristimerin
Molecular Weight 464.64
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

373110R

 
tocris™
RDS373110
10mg Each for $257.85

373150R

 
tocris™
RDS373150
50mg Each for $1,073.10
Description & Specifications

Specifications

Product Type Pristimerin
Inhibitors Pristimerin
Molecular Weight 464.64
Purity >99

Potent and reversible inhibitor of monoacylglycerol lipase (MGL) (IC50 = 93nM). Also suppresses NF-κB activation via inhibition of proteasome chymotrypsin-like activity and IKKαβ. Displays antitumor, anti-inflammatory and antimicrobial activities.

  • Chemical Name: (9β,13α,14β,20α)-3-Hydroxy-9,13-dimethyl-2-oxo-24,25,26-trinoroleana-1(10),3,5,7-tetraen-29-oic acid methyl ester
  • Formula: C30H40O4
  • CAS Number: 1258-84-0
  • Solubility: Soluble to 25mM in DMSO and to 10mM in ethanol
  • Purity: >99