missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Primary Drinking Water Metals without Mercury for Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC EP8N
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ba | 100μg/mL |
Ag | 10μg/mL |
As | 10μg/mL |
Cr | 10μg/mL |
Pb | 10μg/mL |
Cd | 5μg/mL |
Se | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Primary Drinking Water Metals without Mercury | |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental | |
2% HNO3 |
Drinking Water Pollutant Standard | |
125 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Various |
Product intended for laboratory use only