Please login to your online account to display your discounted pricing

PRIMA-1, Tocris

Restores mutant p53 activity; induces apoptosis

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material PRIMA-1
Assay Percent Range >99%
Molecular Formula C9H15NO3
Formula Weight 185.22
Synonym 2, 2-Bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

186250

 
r & d systems
RDS186250
50mg Each for $682.67

186210

 
r & d systems
RDS186210
10mg Each for $164.61
Description & Specifications

Specifications

Formula Weight 185.22
Quantity 50mg
Assay Percent Range >99%
CAS 5608-24-2
Synonym 2, 2-Bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
Molecular Formula C9H15NO3
Recommended Storage Store at +4°C
Chemical Name or Material PRIMA-1
Safety and Handling

Store at +4°C